missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tropylium tetrafluoroborate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Reactant or reagent involved in Retro-Buchner reactions for synthesis of arylcyclopropanes, α-Cyanation of imines, electrochemical redox transformations, oxidative C-H activation of aliphatic aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant or reagent involved in Retro-Buchner reactions for synthesis of arylcyclopropanes, α-Cyanation of imines, electrochemical redox transformations, oxidative C-H activation of aliphatic aldehydes.
Solubility
Soluble in hot methanol.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, strong bases, water/ moisture.
Reactant or reagent involved in Retro-Buchner reactions for synthesis of arylcyclopropanes, α-Cyanation of imines, electrochemical redox transformations, oxidative C-H activation of aliphatic aldehydes.
Solubility
Soluble in hot methanol.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, strong bases, water/ moisture.
Specifications
Specifications
Melting Point | ∼240°C (decomposition) |
Quantity | 5 g |
UN Number | UN1759 |
Beilstein | 3574512 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in hot methanol. |
Formula Weight | 177.94 |
Percent Purity | 97% |
Chemical Name or Material | Tropylium tetrafluoroborate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'