missing translation for 'onlineSavingsMsg'
Learn More

MakerBot™ True Color Large PLA Filament

MakerBot™ True Color Large PLA Filament is composed of the purest grade resin and designed for use with the MakerBot™ 2 Desktop 3D printer and all fifth generation MakerBot™ Replicator 3D printers.

Brand:  MakerBot™ MKS-000004

Additional Details : Weight : 0.90000kg

 View more versions of this product

Product Code. 16301486

  • POA / Each
Estimated Shipment: 17-09-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • Composed of the purest grade resin on the market
  • Comprehensively quality tested
  • Reduces downtime
  • Creates high quality prints
  • Filament Diameter: 1.75 mm
  • Applications: CAD, Conceptual Modeling, Prototyping, Education, AEC
  • Packaging Dimensions (L x W x H): 250 x 250 x 40 mm
Specifications

Specifications

True Green
Rep, Rep Z18, Rep 2, and Rep+ 3D Printers
PLA Filament
PLA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers