missing translation for 'onlineSavingsMsg'
Learn More

MakerBot™ True Color Small PLA Filament

MakerBot™ True Color Small PLA Filament is composed of the purest grade resin and designed for use with the MakerBot™ Replicator and MakerBot™ Replicator Mini 3D printer.

Brand:  MakerBot™ MKS-000057

Additional Details : Weight : 0.35000kg

 View more versions of this product

Product Code. 16361526

  • POA / Each
Estimated Shipment: 17-09-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • Composed of the purest grade resin on the market
  • Comprehensively quality tested
  • Reduces downtime
  • Creates high quality prints
  • Filament Diameter: 1.75 mm
  • Applications: CAD, Conceptual Modeling, Prototyping, Education, AEC
  • Packaging Dimensions (L x W x H): 148 x 148 x 40 mm
Specifications

Specifications

Neon Orange
Rep Z18, Rep 2, Rep Mini, Rep+, and Rep Mini+ 3D Printers
PLA Filament
PLA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers