missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific True North™ Mini-Cooler

The True North™ Mini Cooler protects temperature-sensitive samples at the bench or in the freezer. The Mini Cooler is ideal for keeping PCR™ reagents, enzymes, and other biochemical reagents cool while working at the bench.

Brand:  Heathrow Scientific HS120076

 View more versions of this product

Product Code. 16350686

  • £268.00 / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polycarbonate construction
  • Cooler has rubber non-slip feet
  • Gridded and keyed one-position lid for easy sample identification
  • Available with lid filled with insulating gel
Specifications

Specifications

Blue
4.3 in.
8.9 in.
6.5 in.
Tubes
1 Ea.
Polycarbonate
0.2 mL
11 cm
22.5 cm
16.5 cm
Clear Lid
Tubes
8 x 12
Cooler
-20°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers