missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TrueCut™ Cas9 Protein v2

A next-generation CRISPR Cas9 protein engineered to deliver maximum editing efficiency.

Brand:  Invitrogen™ A36497

 View more versions of this product

Product Code. 15738339

  • £76.75 / 25µg

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Invitrogen TrueCut Cas9 Protein v2 is a next-generation CRISPR Cas9 protein engineered to deliver maximum editing efficiency.

Features include:

  • Consistently high editing efficiency in all tested cell lines including standard, immune, primary, and stem, with up to 2X higher editing efficiency in difficult targets than the competition
  • High quality—manufactured under strict ISO 13485 quality standards
  • Validated protocols for a large number of cell types help you achieve success faster

TrueCut Cas9 Protein v2 is recombinant Streptococcus pyogenes Cas9 (wt) protein, purified from E. coli, for genome editing with CRISPR technology. Cas9 protein forms a very stable ribonucleoprotein (RNP) complex with the guide RNA (gRNA) component of the CRISPR/Cas9 system. Incorporation of nuclear localization signals (NLS) aids delivery to the nucleus, increasing the rate of genomic DNA cleavage.

  • Transfection-ready, using either lipid-mediated transfection reagents or electroporation
  • Eliminates time-consuming cloning steps
  • Available in 2 concentrations:
    • 1 μg/μL for use in standard editing scenarios
    • 5 μg/μL for optimization of editing conditions in more difficult scenarios such as in primary or embryonic cells, microinjection, or when screening multiple gRNA sequences at a time
    • For custom sizes or concentrations, please inquire

Options for obtaining CRISPR gRNA for use with TrueCut Cas9 Protein v2:

  • Order transfection-ready TrueGuide synthetic gRNAs
  • Generate transfection-ready gRNA in as little as four hours including template assembly with our GeneArt Precision gRNA Synthesis Kit (Cat. No. A29377)
  • Have our Custom Services team design, synthesize, and purify in vitro (IVT) gRNA sequences for you

Certifications

CoA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 μg/μL
Molecular Biology
NC_002737.2:854751-858857
901176
cas5, csn1
Untagged
Genome Editing
Cas9 Protein v2
TrueCut™
TrueCut Cas9 Protein v2
TrueCut Cas9 Protein v2 (1 mg/mL), 25 μg

Store at -5°C to -30°C.
E. coli
Spy_1046
Recombinant
Dry Ice
25 μg
Genome Editing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.