Learn More
Invitrogen™ TrueTag™ Donor DNA Kit, RFP stem
The Invitrogen TrueTag Donor DNA Kit, RFP Stem, provides a rapid solution to drive high-efficiency tagging (up to 100% edited cells) of silent or differentially expressed genes with transfection-ready donor DNA.
Brand:  Invitrogen™ A53813
Additional Details : Weight : 0.09000kg
Product Code. 17826511
  • £560.00 / 10 reactions
Estimated Shipment: 20-08-2024
to see stock.
Description
Description
  • A rapid solution to drive high-efficiency tagging (up to 100% edited cells) of silent or differentially expressed genes with transfection-ready donor DNA
  • Kit provides everything needed to produce a high-quality DNA donor to tag gene with a selection marker and GFP Stem
  • Simply provide two locus-specific primers to add homology arms and PCR-amplify the donor
  • One-step PCR amplification to generate donor templates – no cloning required
  • Generate transfection-ready DNA donors in hours instead of days
  • Save up to 3 months with enrichment compared to single cell isolation and characterization
  • Enrich up to 100% RFP Stem-tagged cells
Each kit is designed to enable users to:
  • Enrich tagged cells even when the target protein is not being expressed
  • Choose between a fusion or co-expressed fluorescent tag
  • Choose from puromycin or blasticidin selection markers – whatever best suits the cell model
  • Easily remove antibiotic resistance markers with Cre/Lox following cell enrichment
Included in the kit:
  • Four linear donor templates: C-terminus/puromycin resistance + 2A, C-terminus/blasticidin resistance + 2A, C-terminus/puromycin resistance, C-terminus/blasticidin resistance
  • Phusion Flash High-Fidelity PCR Master Mix
  • PCR cleanup columns and buffers
  • Positive control primers to tag human glial fibrillary acidic protein (GFAP)
Intuitive software to rapidly design gene-specific reagents
Target-specific gene editing reagents are required for proper knock-in of the fluorescent tag. The Invitrogen TrueDesign Genome Editor web-based software does the design work for you:
  1. Identifies target gene transcript
  2. Designates insertion site
  3. Designs and enables easy ordering of all necessary components
Required reagents available via TrueDesign software (not included in kit):
  • CRISPR guide RNA or TALENs designed adjacent to N- or C-terminus
  • Oligos to PCR-amplify your donor and add appropriate homology arms
  • Sequencing primers for assessment of CRISPR-Cas9 cleavage efficiency
  • Verification primers to validate proper insertion of the TrueTag DNA Donor
Also required and available for purchase separately or as part of TrueDesign workflow:
  • TrueCut Cas9 Protein v2, TrueCut HiFi Cas9 Protein, or GeneART CRISPR mRNA
  • CRISPRMAX Transfection Reagent or MessengerMAX Transfection Reagent
Specifications
Specifications
Cellular Imaging
10 reactions
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures