missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trypan Blue Stain Solution, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| CAS | 72-57-1 |
| Molecular Formula | C34H24N6Na4O14S4 |
| MDL Number | MFCD00003969 |
| InChI Key | WMYVHJWZUUEZNE-ARWFNKCKSA-J |
| SMILES | [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1 |
| IUPAC Name | tetrasodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-8-amino-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate |
| Molecular Weight (g/mol) | 960.79 |
| PubChem CID | 9562061 |
| Quantity | 100 mL |
| Chemical Name or Material | Trypan Blue Stain Solution |
Safety and Handling
Product Identifier
- Trypan Blue, 0.4% Solution in PBS
- Danger
- Carcinogenicity Category 1B
- H350-May cause cancer.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : trypan blue
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction