missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TrypLE™ Select Enzyme (10X), no phenol red
GREENER_CHOICE

Product Code. 10698308
Click to view available options
Quantity:
100 mL
500 mL
20 x 100mL
Unit Size:
Each
Pack of 20
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10698308

Brand: Gibco™ A1217703

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

TrypLE™ Select Enzyme (10X), no phenol red

TrypLE™ Select is an animal origin free, recombinant enzyme for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells. We offer a variety of TrypLE™ formulations for a range of cell culture applications.

Compared to trypsin and other dissociation reagents, Gibco™ TrypLE™ Select is:

  • Gentle on cells—maintains cell health for reproducible results
  • Room-temperature stable—convenient to use
  • Easy to use—substitutes directly into existing protocols, with no inactivation required
  • Animal origin free (AOF)—pure, consistent, and effective



    Gentle on cells
    TrypLE™ Select, like trypsin, cleaves peptide bonds on the C-terminal sides of lysine and arginine. However, TrypLE™ Select's exceptional purity increases specificity due to the action of a single enzyme. This reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts.

    Room-temperature stable
    TrypLE™ Select is room-temperature stable and ready to use. TrypLE™ Select cell dissociation reagents remain stable for 24 months at room temperature or 2–8°C, making storage and handling easy and convenient.

    Easy to use
    TrypLE™ Select can be substituted for trypsin in existing protocols. In addition, dilution alone inactivates TrypLE™ Select, avoiding the need for trypsin inhibitors, such as FBS.

    Animal Origin Free (AOF)
    Unlike porcine trypsin and other enzymes, TrypLE™ Select is free of animal-derived components, and is formulated on dedicated animal origin-free equipment.

    Product Intended Use
    For Research Use or Further Manufacturing. Not for use in diagnostic procedures. Customers using Gibco™ TrypLE™ Select in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

    cGMP Manufacturing and Quality System
    Gibco™ TrypLE™ Select is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

    Concentrated forms of TrypLE™ can be aseptically diluted to 1x using a balanced salt solution containing 1 mM EDTA.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Chelators EDTA
    Content And Storage Storage conditions: 15–30°C. Protect from light.
    Alternate storage conditions: 2–8°C or -5 to -20°C.
    Shipping conditions: Ambient.
    Shelf life: 24 months from date of manufacture.
    Classification Animal Origin-free
    Concentration 10 X
    Culture Type Mammalian Cell Culture
    Form Liquid
    Product Type Cell Culture Dissociation Reagent
    Without Additives No Phenol Red
    Green Features Energy efficient, Sustainable packaging
    Inorganic Salts No Calcium, No Magnesium, No Sodium Phosphate
    Osmolality 270 - 320 mOsm/kg
    pH 6.9 to 7.4
    Product Line Gibco
    Purity or Quality Grade Cell Culture Grade
    Quantity 20 x 100mL
    Remarks Room Temperature Stable
    Shipping Condition Room Temperature
    Tested For In vitro Bioassay
    Show More Show Less

    For Research Use or Further Manufacturing. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.