missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TrypLE™ Select Enzyme (1X), no phenol red
GREENER_CHOICE

Gibco™ TrypLE™ Select is an animal origin free, recombinant enzyme for dissociating a wide range of adherent mammalian cells.

Brand:  Gibco™ 12563011

38.09 GBP valid until 2024-12-31
Use promo code "21945" to get your promotional price.


 View more versions of this product

Product Code. 11588846

  • £40.65 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Gibco TrypLE Select is an animal origin free, recombinant enzyme for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells. We offer a variety of TrypLE formulations for a range of cell culture applications.


Compared to trypsin and other dissociation reagents, TrypLE Select is:

Gentle on cells—maintains cell health for reproducible results
Room-temperature stable—convenient to use
Easy to use—substitutes directly into existing protocols with no inactivation required
Animal origin free (AOF)—pure, consistent, and effective

This TrypLE Select is modified as follows:

With
• EDTA

Without
• Phenol Red

Gentle on cells
TrypLE Select, like trypsin, cleaves peptide bonds on the C-terminal sides of lysine and arginine. However, TrypLE Select's exceptional purity increases specificity due to the action of a single enzyme. This reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts.

Room-temperature stable
TrypLE Select is room-temperature stable and ready to use. TrypLE Select cell dissociation reagents remain stable for 24 months at room temperature or 2–8°C, making storage and handling easy and convenient.

Easy to use
TrypLE Select can be substituted for trypsin in existing protocols. In addition, dilution alone inactivates TrypLE Select, avoiding the need for trypsin inhibitors, such as FBS.

Animal Origin Free (AOF)
Unlike porcine trypsin and other enzymes, TrypLE Select is free of animal-derived components and is formulated on dedicated animal origin-free equipment.

Product Intended Use
Customers using TrypLE Select in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP Manufacturing and Quality System
TrypLE Select is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EDTA
Mammalian Cells
1 X
Liquid
No Phenol Red
Bottle(s)
280 - 300 mOsm/kg
Gibco™
Room Temperature Stable
Animal origin-free
Storage conditions: 15–30°C, protect from light.
Alternate storage conditions: 2–8°C or -5 to -20°C.
Shipping conditions: Ambient.
Shelf life: 24 months from date of manufacture.
Animal Origin-free
Adherent Cell Culture
Cell Culture Dissociation Reagent
Low
Energy efficient, Sustainable packaging
7 to 7.4
100 mL
Room Temperature
In vitro Bioassay
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.