missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TrypLE™ Select Enzyme (1X), no phenol red
GREENER_CHOICE

Gibco™ TrypLE™ Select is an animal origin free, recombinant enzyme for dissociating a wide range of adherent mammalian cells.

£41.87 - £148.00

Specifications

Concentration 1 X
Form Liquid
Product Type Cell Culture Dissociation Reagent
Without Additives No Phenol Red
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11588846
GREENER_CHOICE
missing translation for 'viewDocuments'
Gibco™
12563011
100 mL
£41.87
100mL
Please to purchase this item. Need a web account? Register with us today!
11598846
GREENER_CHOICE
missing translation for 'viewDocuments'
Gibco™
12563029
500 mL
£148.00
500mL
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

1 X
Liquid
Cell Culture Dissociation Reagent
No Phenol Red
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months

SDS
Documents

Documents

Certificates

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ TrypLE™ Select Enzyme (1X), no phenol red

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.