Learn More
Gibco™ Trypsin (0.25%), phenol red
Trypsin (0.25%), phenol red
Brand: Gibco™ 25050030
Code : 50
Additional Details : Weight : 0.10000kg
Description
Gibco™ Trypsin solutions are made from trypsin powder—an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength, it is widely used for cell dissociation, during routine cell culture passaging and primary tissue dissociation. We offer a variety of Gibco™ Trypsin and animal origin free TrypLE™ that feature:Quality Testing
Gibco™ Trypsin solutions are tested for pH, osmolality, sterility and performance. In addition, prior to manufacturing, the raw materials are tested for endotoxin, Mycoplasma, bacterial, fungal and viral contamination, as well as multiple activity assays, ash analysis and moisture analysis.
Documented Traceability
We can provide detailed documentation to meet your regulatory needs. Gibco™ Trypsin information available includes lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency.
cGMP Manufacturing and Quality System
Gibco™ Trypsin is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
Specifications
ISO 13485 standards, FDA | |
Trypsin, 20 x 100mL, 1 X Concentration, Liquid, 0.25% Dilution, 9002-07-7 CAS Number, With Phenol Red, No EDTA Chelators, Porcine Pancreas Source, Trypsin Reagent Solution, FDA and ISO 13485 Standards, Used for Mammalian Cell Culture, –5 to –20 | |
1 X | |
Cell Culture Dissociation Reagent | |
No EDTA | |
20 x 100mL | |
Porcine pancreas |
–5 to –20°C | |
Mammalian Cells | |
Liquid | |
Phenol Red | |
Mammalian cell culture | |
For research use only |
Safety and Handling
- Trypsin (1X), liquid
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Trypsin
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.