missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Trypsin/ EDTA Solution
Used to remove cells from the culture vessel surface into suspension. Lonza™Trypsin/ EDTA Solution can be halted by adding a serum media mixture or a Trypsin inhibitor.
Brand: Lonza™ CC-5012
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Clear, colorless-yellow
- Highly active porcine enzyme preparation
- Used at a working concentration of 0.025%
- Prepared in a balanced salt solution, without calcium or magnesium
- pH of 7.6-7.8.
Specifications
Cell Culture Dissociation Reagent | |
100 mL |