missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Trypsin Inhibitor, soybeans

Product Code. p-7082253
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 15443659

Brand: Thermo Scientific Chemicals J60982.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits the pancreatic enzyme trypsin | CAS: 9035-81-8

The soybean trypsin inhibitor has been tested for cell culture applications. It has been used in a study to assess the potential application in animal cell culture of an alkaline protease from a non-toxigenic mangrove isolate of Vibri sp. V26. It has also been used in a study to improve the detection of fungi in eosinophilic mucin. It is an inhibitor of trypsin factor Xa, plasmin and neutrophil elastase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Dissolve in 1X PBS at 1 to 10mg/ml for stock solution
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9035-81-8

Specifications

Quantity 1 g
Description Off-white to Pale Brown
Content And Storage 2°C to 8°C
For Use With (Application) Inhibits the pancreatic enzyme trypsin
Product Type Signal Transduction Reagent
Form Powder
compliance-icons
Product Identifier
  • Trypsin inhibitor, soybeans
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Trypsin inhibitor

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.