missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Trypsin, porcine pancreas
A pancreatic serine protease
Chemical Identifiers
CAS | 9002-07-7 |
---|---|
Molecular Weight (g/mol) | 23.8 kd, 725.80 |
Synonym | EC 3.4.21.4; Trypsin IV porcine |
Content And Storage | -30°C to -10°C |
Form | Lyophilized Powder |
For Use With (Application) | A pancreatic serine protease |
Description | Off-white to Pale Brown |
Description
Trypsin, suitable for use in ELISA or for antigen applications in immunological protocols. Other applications include it is been used for the research purpose.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Reconstitute in aqueous bufferChemical Identifiers
9002-07-7 | |
EC 3.4.21.4; Trypsin IV porcine | |
Lyophilized Powder | |
Off-white to Pale Brown |
23.8 kd, 725.80 | |
-30°C to -10°C | |
A pancreatic serine protease |
Specifications
Off-white to Pale Brown | |
23.8 kd, 725.80 | |
-30°C to -10°C | |
Lyophilized Powder | |
EC 3.4.21.4; Trypsin IV porcine |
A pancreatic serine protease | |
9002-07-7 | |
C35H47N7O10 | |
WGWZNYKOUXOZTC-UHFFFAOYNA-N |
Safety and Handling
missing translation for 'einecsNumber' : 232-650-8
Recommended Storage : Store at -20°C
RUO – Research Use Only