missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptamine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Tryptamine is commonly used in the preparation of biologically active compounds such as neurotransmitters and psychedelics, multi-layered analysis, used to differentiate and identify biogenic amines1
in the synthesis of indole ring-terminated thiol, ω-mercaptooctyltryptamide.This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tryptamine is commonly used in the preparation of biologically active compounds such as neurotransmitters and psychedelics, multi-layered analysis, used to differentiate and identify biogenic amines1 • in the synthesis of indole ring-terminated thiol, ω-mercaptooctyltryptamide.
Solubility
Soluble in DMSO, Methanol. Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Tryptamine is commonly used in the preparation of biologically active compounds such as neurotransmitters and psychedelics, multi-layered analysis, used to differentiate and identify biogenic amines1 • in the synthesis of indole ring-terminated thiol, ω-mercaptooctyltryptamide.
Solubility
Soluble in DMSO, Methanol. Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 111°C to 116°C |
Boiling Point | 137°C (0.15 mmHg) |
Flash Point | 185°C (365°F) |
Quantity | 10 g |
Beilstein | 125513 |
Sensitivity | Air sensitive |
Merck Index | 14,9796 |
Solubility Information | Soluble in DMSO,Methanol. Insoluble in water. |
Formula Weight | 160.22 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'