missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Tryptone Large Granules
Click to view available options
Quantity:
500 g
2000 g
Unit Size:
2kg
500g
Description
- Pancreatic digest of Casein
- Light beige
- Particle size: 1 to 2mm
Specifications
Specifications
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction