Learn More
Thermo Scientific™ Tryptone Soya Agar (Casein Soya Bean Digest Agar)
Description
Grow a wide variety of microorganisms with Thermo Scientific ™ Tryptone Soya Agar. A general purpose agar medium, containing two peptones, Tryptone Soya Agar will support the growth of both aerobes and anaerobes, the latter being grown either in deep cultures or by incubation under anaerobic conditions. Tryptone Soya Agar is recommended as a reference medium when testing selective media to measure the degree of inhibition.
- Has the ready-to-use convenience of prepared media
- May be used as a blood agar base
- Recommended as a reference medium for testing selective media
- Conforms to formulations detailed in International Pharmacopoeia
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2°C to 10°C |
| Format | 90mm Monoplate |
| Media Type | Tryptone Soya Agar |
| Product Type | Agar |
| Fill Volume | 19 mL |
| Quantity | 10/Pk. |
Documents
Product Information
Specification Sheets
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
| Lot Number | Certificate Type | Date | Product Code |
|---|---|---|---|
| 4528447 | Certificate of Analysis | 10/11/2025 | PO0163A |
| 4527326 | Certificate of Analysis | 28/10/2025 | PO0163A |
| 4525562 | Certificate of Analysis | 14/10/2025 | PO0163A |
| 4524513 | Certificate of Analysis | 06/10/2025 | PO0163A |
| 4523727 | Certificate of Analysis | 29/09/2025 | PO0163A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.