missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Tryptone Soya Agar with Disinhibitor PLUS Contact Plate Product Code.: 17631326
  • ONLINE EXCLUSIVE PRICE
    ÂŁ24.30
    ÂŁ22.50 / Pack of 10
    Save ÂŁ1.80
    7% Off

Thermo Scientific™ Tryptone Soya Agar with Disinhibitor PLUS Contact Plate

Product Code. 17631326
TSA with Disinhibitor PLUS, 20/Pk., Germany, Pack of 10
Click to view available options
Description:
TSA with Disinhibitor PLUS
Quantity:
20/Pk.
Manufacturing Origin:
Germany
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17631326

Brand: Thermo Scientific™ PO5172C

  • ONLINE EXCLUSIVE PRICE
    ÂŁ24.30
    ÂŁ22.50 / Pack of 10
    Save ÂŁ1.80
    7% Off
View Available Promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Effortlessly sample surfaces after cleaning and disinfection: four disinhibitors (incl. sodium thiosulfate) neutralize surface disinfectants and aid in microorganism recovery.

Effortlessly sample surfaces for environmental monitoring after cleaning and disinfection using Thermo Scientific™ Tryptone Soy Agar with Disinhibitor PLUS Contact Plates, containing Lecithin, polysorbate 80, sodium thiosulfate and Tween 80 to neutralize surface disinfectants and aid in microorganism recovery. The addition of sodium thiosulfate is in line with International Pharmacopeia requirements.

Streamline environmental monitoring of cleanroom environments with Thermo Scientific™ Tryptone Soy Agar with Disinhibitor PLUS Contact Plates for surface sampling.
  • Secure: dome-shaped bottom prevents agar from falling out of the dish
  • Easy-to-handle: GripFit™ technology ensures the lid remains in place during handling while being easily removed during environmental monitoring
  • Straightforward results interpretation: inner layer on the plate base is sub-divided into grids to allow easy calculation of colony forming units (CFU) from surface tests, enabling labs to report whether contaminant levels fall within pre-defined acceptable limits
  • Easy-to-stack: provides more stability during handling and transport, and ventilation is reduced to prevent plates from drying out
  • Consistent: manufactured using a specialized, automated procedure to ensure a smooth, evenly distributed layer of agar on each plate
Surface sampling is achieved by gently rolling the domed surface of the agar onto the area to be tested. If any microorganisms that can cause disease or infection are present, they will grow to form easily visible colonies on the surface of the agar. This offers a reliable and effective way of determining the microbial cleanliness of surface areas and help control invisible dangers associated with microbial contamination. Clearly visible growth is available within three days for bacteria and within five days for fungi.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Tested in accordance with BP/EP/JP/USP.
Color Ivory, transparent
Content And Storage 2° to 12°C, in the dark
Description TSA with Disinhibitor PLUS
Format 55mm Contact Plate
Manufacturing Origin Germany
Product Type Agar
Fill Volume 13 mL
Fill Weight 13.5g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 20/Pk.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number6335959 Certificate TypeCertificate of Analysis Date06/11/2025 Product CodePO5172C
Lot Number6335960 Certificate TypeCertificate of Analysis Date06/11/2025 Product CodePO5172C
Lot Number6335958 Certificate TypeCertificate of Analysis Date05/11/2025 Product CodePO5172C
Lot Number6333152 Certificate TypeCertificate of Analysis Date29/10/2025 Product CodePO5172C
Lot Number6333153 Certificate TypeCertificate of Analysis Date29/10/2025 Product CodePO5172C
Promo
Expires: 2025-12-31
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.

Applying Promo and Adding...

Promo
Expires: 2025-11-07
Microbiology Madness
Price slashed on over 3000 products

Applying Promo and Adding...

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Tryptone Soya Agar with Disinhibitor PLUS Contact Plate >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.