missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tryptone Soya Agar Irradiated (Triple Wrap) Plate
Carry out environmental monitoring in the most regulated environments with this Triple Wrapped Plate for passive and active air sampling applications.
£41.59 - £77.75
Specifications
Content And Storage | 2 to 25°C |
---|---|
Product Type | Agar |
Packaging Type | Triple Wrapped - Irradiated |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12912618
|
Thermo Scientific™
PO0306T |
Straw, clear |
£77.75
Pack of 25 |
In Stock
Log in to see stock. |
|||||
11965345
|
Thermo Scientific™
PO5012B |
Ivory, transparent |
£41.59
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
Experience next-level cleanroom confidence with Thermo Scientific™ Oxoid™ Tryptone Soya Agar Irradiated Plate. The Tryptone Soya Agar supports the growth of wide variety of organisms.
Perform effortless environmental monitoring with Tryptone Soya Agar Irradiated (Triple Wrap) Plate.
- Sterile: plates are irradiated and triple wrapped in a transparent packaging.
- Consistent: automated manufacturing procedures ensure a smooth, evenly distributed layer of agar on each plate.
- Optimized: products undergo quantitative quality control testing, using a low dose inoculum to ensure optimum performance.
- Ready-to-use: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
2 to 25°C | |
Agar | |
Triple Wrapped - Irradiated |