missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tryptose Phosphate Broth
Cell culture supplement for BHK-21 cells used for vaccine production
Brand: Invitrogen™ 18050039
7.61 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
- Tryptose Phosphate Broth contains tryptose (an enzymatic hydrolysis of protein), dextrose, sodium chloride and disodium phosphate
- Manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK, registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standard
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2 – 8°C Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Tryptose Phosphate Broth | |
100 mL |
Mammalian Cells | |
Sustainable packaging | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.