Learn More
Thermo Scientific™ TSG General Purpose Freezers - Boxed
The TSG Series of general purpose freezers are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.
Brand: Thermo Scientific™ TSG400FESBW
3269.86 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Description
The Thermo Scientific™ TSG Series general purpose laboratory freezers are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.
TSG Series Freezers are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.
- Wide range of sizes from solid door 400 L to 1500 L double door.
- Whisper quite operation < 50 dBA
- White painted interior and exterior
- Adaptive Defrost
- Temperature, door ajar, and power failure (with battery backup) alarms.
- Low heat output
- Rear access port
- Keyed Lock
- LED interior lighting
- Green hydrocarbon refrigerants
- Remote alarm contacts
Specifications
TSG Series General Purpose Laboratory Freezer | |
-12°C to -30°C | |
Solid | |
Steel | |
Painted Steel | |
30 kg | |
Yes | |
1 Rear Access Port | |
Adaptive Defrost | |
493 x 500 x 1405 mm | |
Alarm | |
Smart-Vue Pro Compatible | |
230 V | |
50 Hz | |
R290 |
400 L | |
-20°C | |
1 | |
Painted Steel | |
4 Adjustable | |
CE | |
Digital Microprocessor | |
36 Months | |
Boxed | |
650 x 600 x 1955 mm | |
Key Lock | |
UK | |
230 V/50 Hz | |
Energy Efficient | |
147 kg |