missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12122-1-AP-150UL
This item is not returnable.
View return policy
Description
TSPAN5 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.Specifications
TSPAN5 | |
Polyclonal | |
Unconjugated | |
TSPAN5 | |
NET 4, NET4, Tetraspan NET 4, tetraspanin 5, TM4SF9, TSPAN 5, TSPAN5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10098, 362048, 56224 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62079, P62080, Q68VK5 | |
Tspan5 | |
TSPAN5 Fusion Protein Ag2765 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |