Learn More
TST™ Bowie Dick™ Test Pack
Test the steam penetration and air removal capability of a vacuum sterilizer with this test pack. The TST™ Bowie Dick Test Pack is a unique combination of advance Browne intelligent ink technology and the original Bowie Dick concept.
Brand:  Steris Corporation 2352AB
Code : 33
Additional Details : Weight : 10.80000kg
 View more versions of this product
Product Code. 12380489
  • £449.95 / Pack of 20
In Stock
to see stock.
Add to basket
Description
Description
  • Single-use test pack
  • A successful test confirms that steam penetration into a test pack is rapid and even and that air and other non-condensable gases have been effectively removed
  • The chemical indicator sheet at the center of the pack shows a defined color change from yellow to dark blue/purple when exposed to a specific combination of time, temperature and steam
  • Conforms to ISO 11140-4 Class 2
Specifications
Specifications
Autoclavable
For large porous load sterilizers
TST Bowie Dick Test Pack
134 to 137˚C for up to 3.5 min., Conforms to ISO 11140-4(3) Class 2
Yes
Steam
ISO 11140-4(3) Class 2
Autoclave
134°C to 137°C
100 Pk.
3.5 min.