missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Freezer Exterior Illumination Accessories

This TSX freezer exterior illumination option provides sample safety alert and alarm indications to protect your samples.

£348.50 - £436.51

Specifications

Cabinet Material Plastic
Color Clear
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Length (Metric) Exterior Length (English) Exterior Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Length (Metric) Exterior Length (English) Exterior Price Quantity & Availability  
17344650
View Documents
Thermo Scientific™
TSXLBLG
For use with TSX600/700 Freezers 920 mm 36 in.
£436.51
Each
In Stock
to see stock.
Add to basket
 
17354650
View Documents
Thermo Scientific™
TSXLBSM
For use with TSX400/500 Freezers 500 mm 20 in.
£348.50
Each
In Stock
to see stock.
Add to basket
 
Description

Description

This TSX freezer exterior illumination option provides sample safety alert and alarm indications to protect your samples

Specifications

Specifications

Plastic
Clear
Videos
SDS
Documents

Documents

Product Certifications

Only compatible with TSX units built after May 25, 2021. See label on left side of TSX cabinet for manufacturing date.