missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Series High-Performance Undercounter Lab Refrigerators, Glass Door, VV

For a better environment inside and out - Introducing our new undercounter lab refrigerator.

Brand:  Thermo Scientific™ TSX505GV

Additional Details : Weight : 75.00000kg

Product Code. 15713019

  • £4820.00 / Each
Estimated Shipment: 19-12-2024
to see stock.

Description

Description

Designed for your environment, new Thermo Scientific™ TSX Series high-performance undercounter lab refrigerators support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other research and medical products. Powered by V-drive, the TSX Series delivers low energy, decreased noise levels and improved performance and user experience. These new undercounter refrigerators are quiet and feature an improved user interface, automated locking and increased storage capacity, and reliability.

Designed to maintain temperature uniformity to protect medical, pharmaceutical-grade products, laboratory media, reagents and more. 
  • Unique V-drive technology is designed to save energy without compromising the protection of stored products
  • U.S. Environmental Protection Agency SNAP (Significant New Alternatives Policy) compliant refrigerants
  • Quiet operation at just 35dbA (43 during pull down)
  • Reduced energy consumption
  • Sleek exterior design and touch screen
  • Bright, easy-to-adjust interior LED lighting
  • Touchscreen controller with password protection
  • Datalogging with USB download functionality
  • Battery backup
  • Choice of reversible glass or solid door
  • Air or glycol temperature display
  • Automated locking
  • 1" rear access port
  • Remote alarm contacts
  • Audible and visual alarms: power fail, high temp, low temp, low battery, memory full, door ajar
  • ADA compliant
  • Compatible with most major electronic locks*

*Data on file.
Specifications

Specifications

TSX Series High-Performance Undercounter Refrigerator, Glass Door
5.5 cu. ft.
3°C to 7°C
Single Glass
3
Undercounter
TSX Series
Alarm
20.5 x 19.5 x 23 in.(52 x 49.5 x 58.4 cm)
26 in.
66 cm
31.8 in.
79.5 cm
23.6 in.
60 cm
10 A Max / 3 A Steady State
100/240 V
50/60 Hz
155 lb.
119 lb.
High-Performance Refrigerator
156 L
R744
1
CE, REACH, RoHS, UL
Full color TFT-LCD; 4.3”; 480 x 272
Automatic
Smart-Vue Compatible
26 x 23.6 x 31.8 in. (66 x 60 x 81 cm)
20.5 in.
52 cm
23 in.
58.4 cm
19.5 in.
49.5 cm
EU
100 - 240 V 50-60 Hz
Energy efficient
71 kg
53.98 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Services