missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Series High-Performance Chromatography Refrigerators
GREENER_CHOICEBIOPHARMA

Innovative refrigeration solutions for healthcare and research needs

£6513.89 - £9153.99

Specifications

Refrigerant R290
Display Microprocessor
Product Line TSX Series
Defrost Automatic
Monitoring Options Alarm, Recorder
View More Specs

Products 4
Product Code Brand Plug Type Door Style Electrical Requirements Price Quantity & Availability  
Product Code Brand Plug Type Door Style Electrical Requirements Price Quantity & Availability  
16367181
BIOPHARMA
View Documents Promotion Details
Thermo Scientific™
TSX2305CV
CEE 7/7 Single Glass 208-230V, 50/60Hz
£6513.89
Each
Please to purchase this item. Need a web account? Register with us today!
16243952
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Thermo Scientific™
TSX4505CV
CEE 7/7 Sliding Glass 208-230V, 50Hz
£9153.99
Each
Please to purchase this item. Need a web account? Register with us today!
16293442
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Thermo Scientific™
TSX5005CV
CEE 7/7 Double Glass 208-230V, 50Hz
£9153.99
Each
Estimated Shipment: 25-03-2025
to see stock.
Add to basket
 
16387181
BIOPHARMA
View Documents Promotion Details
Thermo Scientific™
TSX3005CV
CEE 7/7 Glass 208-230V, 50Hz
£7116.23
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific™ TSX Series high-performance chromatography refrigerators are designed for a variety of applications requiring precise temperature control, full access to chromatography instrumentation, and easy setup of instrumentation and apparatus within the chamber. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns and offers significant energy savings without compromising protection.

The TSX Chromatography series features industry-leading safety features to ensure maximum protection as well as delivering over 450W of reserve capacity to handle the heat load generated by instruments within the cabinet and to react quickly to frequent door access.

The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

The drive for sample integrity and security
  • Designed to safely store chromatography and other lab instrumentation requiring a temperature range of 2°C to 8°C
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance
  • Two 2" (5 cm) side wall access ports with insulated hinged covers for solvent, waste, and other common tubing and hoses used in chromatography applications.
  • Hydrocarbon Safety switch protected internal Duplex outlet standard on all models 
 
Enabling compliance
  • Tested to GMP Clean Room Class A / ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
  • Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863
 
Designed for an ergonomic and comfortable experience
  • Ergonomically designed door handle with a soft-touch grip
  • The controller is angled to help prevent neck strain
  • Convenient, easy to clean and adjustable epoxy coated full and half shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Bright, LED interior lighting
  • Self-closing glass doors with 90° stop to assist with inventory loads
 
Verified sustainability
  • US EPA ENERGY STAR Certified with up to 40% less energy usage than conventional refrigerant models*
  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life. **
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility
 
Ready for connectivity and monitoring
  • Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always—and anywhere— know the safety of your samples and the performance of your refrigerator or freezer.
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
 
Make it your own
  • Alternate Door swing configuration and door types
  • Extra and Alternate wire baskets, shelves, and drawers
  • Optional Chromatography support shelves
  • Free-standing or factory-installed chart recorders
  • Stainless Steel interior finish
  • E-Lock Adapter kits 
  • 4-20 milliamp transmitters and other remote alarm modules
  • Specialty, regional, and country specific plugs
 
Services and support
  • Industry Leading 120-month compressor warranty (varies by country, please contact us for details)
  • Optional Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate
  • Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
  • Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs
 
Specifications

Specifications

R290
TSX Series
Alarm, Recorder
Smart-Vue Compatible
Microprocessor
Automatic
Optional
78.5 in.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.