Learn More
Jaytec Glass™ Gas Sampling Tube
The Gas Sampling Tube is manufactured in accordance with BS.2069.
Brand:  Jaytec Glass™ GRF246
Code : 02
Additional Details : Weight : 0.01000kg
Product Code. 12308597
  • £77.65 / Each
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
Description
Description
  • Borosilicate glass
  • One-way glass stopcock
Specifications
Specifications
250 mL
Borosilicate Glass
Gas Sampling Tube