missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Tube Holder

OHAUS™ Tube Holder is constructed of polyethylene foam and can be used in a range of applications.

Brand:  OHAUS™ 30400230

26.79 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.



Additional Details : Weight : 0.90000kg

 View more versions of this product

Product Code. 15722649

  • £28.20 / Pack of 2
Estimated Shipment: 08-11-2024
to see stock.
Add to basket

Description

Description

  • Can be used on all vortex mixers
  • Increase throughput by mixing multiple samples at one time
  • Constructed of polyethylene foam
  • Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications

Specifications

14 to 19 mm Tubes
Tube Holder
Holder For 14-19 mm Tubes, Pack of 2
2/Pk.
14 to 19 mm
Foam
8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers