Learn More
OHAUS™ Tube Holder
OHAUS™ Tube Holder is constructed of polyethylene foam and can be used in a range of applications.
Brand: OHAUS™ 30400232
57.86 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.90000kg
Description
- Can be used on all vortex mixers
- Increase throughput by mixing multiple samples at one time
- Constructed of polyethylene foam
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Microtubes | |
Foam | |
48 |
Tube Holder | |
Micro-Tube Holder, Pack of 2 | |
2/Pk. |