Learn More
OHAUS™ Tube Holder
OHAUS™ Tube Holder is constructed of polyethylene foam and can be used in a range of applications.
Brand:  OHAUS™ 30400204
Additional Details : Weight : 0.90000kg
 View more versions of this product
Product Code. 15762619
  • £65.70 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Can be used on all vortex mixers
  • Increase throughput by mixing multiple samples at one time
  • Constructed of polyethylene foam
  • Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Specifications
18 x 0.5 mL Microtubes
Foam
18
Tube Holder
0.5 mL Micro-Tube Holder