missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Tube Holder

OHAUS™ Tube Holder is constructed of polyethylene foam and can be used in a range of applications.

Brand:  OHAUS™ 30400205

57.16 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.



Additional Details : Weight : 0.90000kg

 View more versions of this product

Product Code. 15772619

  • £65.70 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Can be used on all vortex mixers
  • Increase throughput by mixing multiple samples at one time
  • Constructed of polyethylene foam
  • Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications

Specifications

24 x 1.5 to 2 mL Microtubes
Foam
24
Tube Holder
1.5-2.0 mL Micro-Tube Holder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers