Learn More
Cytiva Tube Holder AKTA™ avant polyoxymethylene
Brand: Cytiva 28954329
This item is not returnable.
View return policy
Description
For the organisation of small tubing on the wet side of ÅKTA avant or ÅKTA pure systems. Attached to the system by small holes along left edge of wet side and on pump fronts.
Specifications
Tube Holder | |
Organize small tubing on the wet side of AKTA avant or AKTA pure | |
Polyoxymethylene (POM-C) | |
Pk. of 5 |
Product Content Correction
Product Title
Cytiva Tube Holder AKTA™ avant polyoxymethylene > Tube Holder AKTA avant polyoxymethylene
Spot an opportunity for improvement?Share a Content Correction