Learn More
Greiner Bio-One Tube, Polypropylene, Round Bottom, Non-Sterile
Suitable for a variety of purposes and featuring good thermal, chemical, and mechanical stability.
Brand:  Greiner Bio-One 115201
Product Code. 10353821
  • £120.00 / Pack of 2000
Explore more special offers
Description
Description
  • Polypropylene tubes have particularly good thermal, chemical and mechanical stability
  • Material is ideally suited for the storage of chemical and biological samples
TRUSTED_SUSTAINABILITY
Specifications
Specifications
12 mm
Polypropylene
250/Pk.
5 mL
75 mm
Non-sterile