Learn More
Fisherbrand™ Tube Roller Rack
Efficient and practical solution for tube roller workflows.
Brand: Fisherbrand™
Additional Details : Weight : 0.20000kg
Description
Expand capacity of your workflow. Each Fisherbrand Tube Roller Rack accommodates 10 tubes, doubling or tripling the capacity of your tube roller equipment.
- Saves Process Time - Large batch processing reduces the number of roller cycles required for lab procedures
- Saves Money - Handles more without the need to invest in multiple machines to process the same amount of tubes
- Saves Valuable Space - Additional capacity especially helpful with limited incubator capacity
- Precise and efficient tube rolling
- Tube holder axis is parallel to roller axis for pure horizontal rotation for controlled sample manipulation.
- Provides a Gentler Mix -- Tube Roller Rack slows the effective RPM of the tube providing a slower mix (New RPM Calculation = [Speed Value] * 0.26)
- Preserves Sample Integrity -- Controlled, lower speed mitigates cell damage reducing sample errors such as hemolysis
- Organize samples pre and post mixing
- Use as a storage tube rack for upright storage or submerse in an ice bath
- Polypropylene construction is wear-resistant and offers reliable sterilization through autoclaving
- Silicone rings for smooth rotation and gentle contact with roller
Specifications
10 Tubes | |
Tube Roller Rack, Small | |
0.6 lb. | |
Blue | |
Pack of 3 Racks |
3.5 to 7 mL | |
Polypropylene, Silicone | |
0.27 kg | |
Holding transport, conical bottom, blood collection and culture tubes (3.5 to 7 mL) up to 10 to 17 mm |