Learn More
Sartorius Tubed Media for Sartorius Membrane Filtration Units: Jus de Tomate Agar
Tubed media provides time and cost savings
Brand: Sartorius 14140----------K
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The traditional culture media for microorganisms is agar media. This can be used for the membrane filtration method or for direct incubation. Agar media in tubes are for pouring agar plates. The content of one tube is sufficient for two 90mm or three 60mm Petri dishes.
- 20mL tubes, 50 tubes per box
- According to international formulas
- Consistent quality
Specifications
Liquid | |
Jus de Tomate Agar | |
Membrane filtration method or for direct incubation, for determination of Leuconostoc oenos and other wine spoiling organ | |
20 mL |
Germany | |
20 mL | |
Bottle |
Safety and Handling
- X50 20ML MEDIA JUS TOMATE ST
Signal Word
- Danger
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose