Learn More
Sartorius Tubed Media for Sartorius Membrane Filtration Units: Nutrient Agar
Tubed media provides time and cost savings
Brand: Sartorius 14137----------K
Code : NEW
Additional Details : Weight : 0.50000kg
Description
The traditional culture media for microorganisms is agar media. This can be used for the membrane filtration method or for direct incubation. Agar media in tubes are for pouring agar plates. The content of one tube is sufficient for two 90mm or three 60mm Petri dishes.
- 20mL tubes, 50 tubes per box
- According to international formulas
- Consistent quality
Specifications
Liquid | |
Nutrient Agar | |
Membrane filtration method or for direct incubation, for determination of total count | |
20 mL |
Germany | |
20 mL | |
Bottle |