Learn More
Thermo Scientific™ Tubes and Domed Caps, strips of 8
Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.
£159.00 - £283.00
Specifications
Certifications/Compliance | Certified DNA/RNase Free |
---|---|
Color | Natural |
Volume (Metric) Working | 0.2 mL |
Volume (Metric) | 0.25 mL |
For Use With (Equipment) | Thermal Cyclers |
Product Code | Brand | Color | Product Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Product Type | Quantity | Price | Quantity & Availability | |||
10249393
|
Thermo Scientific™
AB0266 |
Natural | 8-Strip Tube | 250 Strips |
£283.00
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10475353
|
Thermo Scientific™
AB0451 |
Natural | 8-Strip Tube | 10 Packs of 12 Strips |
£159.00
Pack of 120 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.
Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.
Highlights- Compatible with 0.2 mL thermal cycler blocks 8 tubes per strip 0.25 mL maximum tube capacity when capped
- Sealable using flat and domed cap strips
- Ultra-Clear cap options available for use in qPCR assays
- PCR
- qPRC
- Real-Time PCR
Specifications
Certified DNA/RNase Free | |
0.2 mL | |
Thermal Cyclers | |
Standard | |
96 |
Natural | |
0.25 mL | |
Polypropylene | |
Domed Cap | |
8-Strip Tube |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.