missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palintest™ Tubetest COD Reagent Tube with Mercury For High Chloride Sample
Suitable for working with samples with very high chloride levels, this high chloride formulation (2M), is ideal for working in laboratory and other types of scientific environments.
£138.00 - £169.00
Specifications
For Use With (Application) | High Chloride Samples Such as Chemical Oxygen Demand (COD) |
---|---|
For Use With (Equipment) | Photometer or UV-Vis Spectrophotometer |
Includes | Chemical Oxygen Demand (COD) |
No. of Tests | 25 |
Type | COD Reagent Tube |
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
11918937
|
Palintest™
PL461 |
0 to 150mg/L |
£169.00
Pack of 24 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11938937
|
Palintest™
PL467 |
0 to 20000mg/L |
£138.00
Pack of 24 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The mercury free formulation is recommended for general use
- The mercuric sulfate formulation (M) is provided for use in standard procedures which require the suppression of interference due to moderate levels of chloride
- The high chloride formulation (2M) provides much higher levels of chloride suppression and is suitable for working with samples with very high chloride levels
- The new M/C formulation with mercuric sulfate is specifically for use in non-Palintest™ instruments
Specifications
High Chloride Samples Such as Chemical Oxygen Demand (COD) | |
Chemical Oxygen Demand (COD) | |
COD Reagent Tube |
Photometer or UV-Vis Spectrophotometer | |
25 |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P314-Get medical advice/attention if you feel unwell.
- P321-Specific treatment (see on this label).
- P363-Wash contaminated clothing before reuse.
- P391-Collect spillage.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title