missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBGCP3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15719-1-AP-20UL
This item is not returnable.
View return policy
Description
15719-1-AP antibody detects the isoform proteins around 95-101 and 50 kDa in SDS-PAGE.
The gamma-Tubulin complex is composed of gamma Tubulin and the gamma-Tubulin complex associated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. gamma-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the gamma-Tubulin complex. GCP3 (gamma-tubulin complex component 3), also known as TUBGCP3 or SPBC98, localizes to the centrosome and is an ubiquitously expressed 907 amino acid member of the gamma-Tubulin complex.Specifications
TUBGCP3 | |
Polyclonal | |
Unconjugated | |
TUBGCP3 | |
GCP 3, GCP3, h104p, hGCP3, hGrip104, hSpc98, SPBC98, Spc98p, TUBGCP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10426 | |
-20°C | |
Liquid |
Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96CW5 | |
TUBGCP3 | |
TUBGCP3 Fusion Protein Ag8437 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction