missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tubulin Tracker™ Deep Red

Tubulin Tracker™ Deep Red provides deep-red/far-red fluorescence when bound to polymerized tubulin in live cells.

Brand:  Invitrogen™ T34076

341.24 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 16180561

  • £449.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Based on Docetaxel conjugated with a bright, photostable deep-red fluorophore
  • Docetaxel belongs to the family of cytoskeletal drugs that target tubulin
  • Absorbs and emits optimally at 652 nm and 669 nm, respectively and can be visualized with standard Cy5 filter settings using almost any fluorescent imaging instrument
  • Can be multiplexed with blue, green, orange, red, and near-IR fluorophores.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains sufficient material for at least 300 slides at 1 mL per slide.

  • 5 vials Tubulin Tracker Deep Red (Add 60 μL/vial of anhydrous DMSO to make a 1000X (1mM) stock solution.  Dissolve stock solution 1:1000 in cell imaging solution to make 1X (1 μM) staining solution.)
    • 5 x 77 mg Probenecid (Dissolve vial contents in 1 mL of live cell compatible buffer to make a 100X stock solution.)

      Store at ≤–20°C after recieving.
      Desiccate and protect from light.
      When stored as directed, product is stable for at least 6 months.

300 slides
Approved for shipment on Wet or Dry Ice
Tubulin Tracker
652⁄669
TubulinTracker™
UV-Vis Labels
Tubulin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.