missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tubulin Tracker™ Variety Pack

Product Code. 15913322
Click to view available options
Quantity:
2 x 60 Slides
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15913322

Brand: Invitrogen™ T34079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Tubulin Tracker Variety Pack is a kit containing Tubulin Tracker Green and Tubulin Tracker Deep Red, which provide green and deep-red fluorescence, respectively, when bound to polymerized tubulin in live cells.

Tubulin Tracker Green provides fluorescence with excitation maxima of 494 nm and emission maxima of 522 nm and can be detected with standard FITC/GFP filter settings using a fluorescence microscope. Tubulin Tracker Deep Red provides fluorescence with excitation maxima of 652 nm and emission maxima of 669 nm and can be detected using standard Cy5 filter settings.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains sufficient material for at least 60 slides each for Tubulin Tracker Green and Tubulin Tracker Deep Red at 1 mL / slide and 1 x working concentration.This kit contains one Tubulin Tracker Green kit (Cat. No. T34078) and one Tubulin Tracker
Excitation Wavelength Range Tubulin Tracker Deep Red: 652/669, Tubulin Tracker Green: 494/522
Quantity 2 x 60 Slides
Product Line TubulinTracker
Shipping Condition Approved for shipment on Wet or Dry Ice
Label Type UV-Vis Labels
Product Type Tubulin Tracker
SubCellular Localization Tubulin

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.