missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ Tumor Antigen Microarray

Tumor Antigen Microarray with 4319 antigen specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

Brand:  PEPperCHIP™ SPC.023.001

Product Code. 16357768

  • £515.00 / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Microarray Content: histone H1.2, histone H4, G2/mitotic-specific cyclin-B1, L-dopachrome tautomerase, p53, receptor tyrosine-protein kinase erbB-2 , mammaglobin A, TCRgamma alternate reading frame protein, stromelysin-3, protein SSX2, prostate-specific antigen, cancer/testis antigen 1, myc proto-oncogene, claudin-6, carcinoembryonic antigen-related cell adhesion molecule 5, Melanocyte protein PMEL, Wilms tumor protein , tyrosinase, SOX-2, myelin-oligodendrocyte glycoprotein, actin, alpha skeletal muscle , HLA classI, A-36 alpha chain
  • Peptide Length/Overlap: 15 aa/ 13 aa
  • Number of Peptides: 4319 printed in duplicate and HA, polio and c-Myc control peptides
Specifications

Specifications

PEGMA/PMMA
1 Slide
1 mm
Transparent
Individual barcode for each microarray
High-density Peptide Microarray
Non-sterile
1 array copy per microarray
Combinatorial peptide synthesis by amino acid laser printing
ISO 9001:2015
RUO
Microscopic Glass
Antibody validation, epitope mapping, serum screening, biomarker discovery
75.4 x 25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.