Learn More
Tungsten boride, 99.4% (metals basis), Thermo Scientific Chemicals
£49.85 - £203.01
Chemical Identifiers
CAS | 12007-98-6 |
---|---|
Molecular Formula | B5W2 |
Molecular Weight (g/mol) | 421.73 |
MDL Number | MFCD00049707 |
InChI Key | ZFRNSGNJMJAUMG-UHFFFAOYSA-N |
Synonym | ditungsten pentaboride, w |
PubChem CID | 25113386 |
IUPAC Name | boranylidyne($l^{2}-boranylidene)tungsten;boranylidynetungsten;$l^{2}-boranylideneboron |
SMILES | [B]=[B].B#[W].B#[W]=[B] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358808
|
Thermo Scientific Alfa Aesar
041664.14 |
25 g |
£49.85
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368808
|
Thermo Scientific Alfa Aesar
041664.22 |
100 g |
£203.01
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12007-98-6 | |
421.73 | |
ZFRNSGNJMJAUMG-UHFFFAOYSA-N | |
25113386 | |
[B]=[B].B#[W].B#[W]=[B] |
Specifications
12007-98-6 | |
B5W2 | |
ditungsten pentaboride, w |
|
ZFRNSGNJMJAUMG-UHFFFAOYSA-N | |
boranylidyne($l^{2}-boranylidene)tungsten;boranylidynetungsten;$l^{2}-boranylideneboron | |
25113386 | |
99.4% | |
(metals basis) |
Powder | |
MFCD00049707 | |
Insoluble in water. | |
[B]=[B].B#[W].B#[W]=[B] | |
421.73 | |
421.76 | |
Odorless | |
Tungsten boride |
Safety and Handling
missing translation for 'einecsNumber' : 234-524-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.