missing translation for 'onlineSavingsMsg'
Learn More

Tungsten boride, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11337407
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

it is used as an abrasives used in thin, thick Film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-09-9
Molecular Formula BW
Molecular Weight (g/mol) 194.65
MDL Number MFCD00049708
InChI Key JEEHQNXCPARQJS-UHFFFAOYSA-N
Synonym tungsten boride, tungsten monoboride, tungstenioborane
PubChem CID 82793
IUPAC Name boranylidynetungsten
SMILES B#[W]

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information It is insoluble in water.
Formula Weight 194.66
Percent Purity 99%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Tungsten boride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.