missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Tungsten Bulb Filament

Tungsten halogen lamp with horizontal filament for Ultrospec 500 Pro UV/Visible Spectrophotometer.

Brand:  Biochrom 80-2022-94

Code : GE

Additional Details : Weight : 0.00700kg

Product Code. 10794787

  • £84.35 / Each
Estimated Shipment: 06-09-2024
to see stock.

Description

Description

  • Tungsten halogen lamp
  • Horizontal filament
  • For use with Ultrospec 500 Pro UV/Visible spectrophotometer
Specifications

Specifications

Halogen Lamp
Tungsten Halogen
20 W
Vertical
Wire Terminal
12 V
UV and Visible
Ultrospec 500 Pro UV/Visible Spectrophotometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers