missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten carbide Cobalt, 99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
Description
Tungsten carbide Cobalt is used in cutting tools, having hard ceramic crystals of a few microns in size within a ductile metal matrix, use as electrodes for Electrode Discharge Machining. It is also used in production of tools for milling, drilling and pressing/punching processes, surgical instruments, product is the rotating balls used in ball point pens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Gray |
Avg. Mol. Wt. or Mol. Wt. Range | WC:Co; 94:6 wt% |
UN Number | UN3178 |
Quantity | 1 kg |
Solubility Information | Insoluble in water. |
IUPAC Name | methylidynetungsten cobalt |
Percent Purity | 99% |
Mesh Size | −325 Mesh |
Odor | Odorless |
Form | Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'