missing translation for 'onlineSavingsMsg'
Learn More
Tungsten dichloride dioxide, 99%, Thermo Scientific Chemicals Product Code.: 11473870

Tungsten dichloride dioxide, 99%, Thermo Scientific Chemicals

Product Code. 11473870
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11473870

Brand: Thermo Scientific Alfa Aesar 089962.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tungsten dichloride dioxide is used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13520-76-8
Molecular Formula WO2Cl2
MDL Number MFCD00054136

Specifications

Color Yellow
Physical Form Powder
Quantity 50 g
Assay Percent Range 99%
Solubility Information Soluble in cold water. Decomposes in hot water.
Formula Weight 286.75
Odor Acrid
Sensitivity Moisture sensitive
Chemical Name or Material Tungsten dichloride dioxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten dichloride dioxide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.