missing translation for 'onlineSavingsMsg'
Learn More
Tungsten foil, 0.25mm (0.01in) thick, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11335918

Tungsten foil, 0.25mm (0.01in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11335918
100 x 100 mm
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
This item is not returnable. View return policy

Product Code. 11335918

Brand: Thermo Scientific Alfa Aesar 010415.GH

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx. 12g/50 x 50mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powderShow MoreShow Less
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Assay Percent Range 99.95% (Metals basis)
Quantity 100 x 100 mm
Percent Purity 99.95%
Weight ≈12g/50x50mm
Form Foil
Assay (metals basis)
Chemical Name or Material Tungsten foil, 0.25mm (0.01 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU23K014 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number010415.GH,010415.FI,010415.HP
Lot NumberZ27J022 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number010415.GH,010415.FI,010415.HP
Lot NumberU08J027 Certificate TypeCertificate of Analysis Date21/08/2023 Catalog Number010415.GH,010415.FI,010415.HP
Lot NumberQ28I054 Certificate TypeCertificate of Analysis Date23/11/2022 Catalog Number010415.GH,010415.FI,010415.HP
Lot NumberQ15I010 Certificate TypeCertificate of Analysis Date22/04/2022 Catalog Number010415.GH,010415.FI,010415.HP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten foil, 0.25mm (0.01in) thick, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.