missing translation for 'onlineSavingsMsg'
Learn More
Tungsten gauze, 150 mesh woven from 0.02mm (0.0008in) dia wire, Thermo Scientific Chemicals Product Code.: 15427435

Tungsten gauze, 150 mesh woven from 0.02mm (0.0008in) dia wire, Thermo Scientific Chemicals

Product Code. 15427435
75 x 75 mm
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
This item is not returnable. View return policy

Product Code. 15427435

Brand: Thermo Scientific Alfa Aesar 047167.FL

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 75 x 75 mm
Mesh Size 150 Mesh
Form Wire Cloth
Chemical Name or Material Tungsten gauze, 150 mesh woven from 0.02mm (0.0008 in.) dia. wire

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN28L012 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog Number047167.GT,047167.FL
Lot NumberZ20H009 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number047167.GT,047167.FL,047170.06
Lot NumberR21H022 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number047167.GT,047167.FL,047170.06
Lot NumberT28G022 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number047167.GT,047167.FL,047170.06
Lot NumberP19F045 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number047167.GT,047167.FL,047170.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten gauze, 150 mesh woven from 0.02mm (0.0008in) dia wire, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.