Learn More
Tungsten ICP Standard, 1000 mg/L W in NH3 4%, Honeywell Fluka™
Brand: Honeywell Fluka E1127-100ML
Additional Details : Weight : 0.10000kg
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Tungsten ICP Standard | |
Standards for ICP and ICP-MS | |
>11.5 | |
100 mL |